Magnesium ≥99.98% (metals basis), turnings -4 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
36193-18
36193-30
CAAA36193-18EA
68.73
CAD
CAAA36193-18
CAAA36193-30
Magnesium ≥99.98% (metals basis), turnings -4 mesh
Magnesium
For vacuum deposition
Purity: 99.98%
CAS No.: 7439-95-4
Odor: Odorless
UN No.: 1869
MDL No.: MFCD00085308
For vacuum deposition
Formula:
Mg MW: 24.3 g/mol Boiling Pt: 1107 Melting Pt: 651 Storage Temperature: Ambient |
MDL Number:
MFCD00085308 CAS Number: 7439-95-4 EINECS: 231-104-6 UN: 1869 ADR: 4.1 III Merck Index: 13,05675 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...