Sodium selenide ≥99.8% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

36187-03 36187-06
CAAA36187-03EA 630.42 CAD
CAAA36187-03 CAAA36187-06
Sodium selenide ≥99.8% (metals basis)
Sodium selenide

MDL: MFCD00014240


Purity: 99.8% CAS No.: 1313-85-5 Molecular Formula: Na2Se Formula Weight: 124.94 Physical Form: -60 Mesh (dry under argon) Density: 2.625 Appearance: Grey to pink Melting Point: 875° Merck Reference: 14,8672 UN No.: 3283 MDL No.: MFCD00014240

Formula: Na₂Se
Melting Pt: >875 °C
Density: 2.625
Storage Temperature: Ambient
MDL Number: MFCD00014240
CAS Number: 1313-85-5
EINECS: 215-212-0
UN: 3283
ADR: 6.1,II
Merck Index: 14,8672

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR