Boron tribromide ≥99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
35846-18
35846-30
CAAA35846-18EA
178.54
CAD
CAAA35846-18
CAAA35846-30
Boron tribromide ≥99%
Boron tribromide
As a catalyst in the manufacturing of diborane, ultra pure boron
Purity: 99% min
CAS No.: 10294-33-4
Molecular Formula: BBr3
Formula Weight: 250.52
Physical Form: Liquid, ampouled under argon
Density: 2.643
Appearance: Colorless to pale orange
Melting Point: -46°
Boiling Point: 90°
Merck Reference: 14,1347
UN No.: 2692
MDL No.: MFCD00011312
As a catalyst in the manufacturing of diborane, ultra pure boron
Formula:
BBr₃ MW: 250.52 g/mol Boiling Pt: 91.2 Melting Pt: -46 Density: 2.643 Storage Temperature: Ambient |
MDL Number:
MFCD00011312 CAS Number: 10294-33-4 EINECS: 233-657-9 UN: 2692 ADR: 8 I Merck Index: 14,1347 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...