Bismuth (III) sulfide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dibismuth trisulphide, Dibismuth trisulfide

35834-18 35834-30
CAAA35834-18EA 221.2 CAD
CAAA35834-18 CAAA35834-30
Bismuth (III) sulfide ≥99.9% (metals basis)
Bismuth (III) sulphide

MDL: MFCD00014204


Purity: 99.9% CAS No.: 1345-07-9 Molecular Formula: Bi2S3 Formula Weight: 514.15 Physical Form: Chunks/Crystals Density: 7.7 Appearance: Brown/black MDL No.: MFCD00014204

Formula: Bi₂S₃
Melting Pt: 763 °C
Density: 7.7
Storage Temperature: Ambient
MDL Number: MFCD00014204
CAS Number: 1345-07-9
EINECS: 215-716-0
Merck Index: 12,01329

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR