Copper ≥99.8% (metals basis), thin foil, Thickness: 0.01 mm (0.0004 in)
Supplier: Thermo Scientific Chemicals
35815-FF
35815-FI
35815-GH
CAAA35815-FFEA
96.22
CAD
CAAA35815-FF
CAAA35815-FI
CAAA35815-GH
Copper ≥99.8% (metals basis), thin foil, Thickness: 0.01 mm (0.0004 in)
Copper
Purity: 99.8%
CAS No.: 7440-50-8
Physical Form: ≈0.06g/25x25mm, not certified pinhole free
Odor: Odorless
MDL No.: MFCD00010965
Formula:
Cu MW: 63.55 g/mol Boiling Pt: 2595 Melting Pt: 1083 Density: 8.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...