Titanium ≥99.6% (metals basis), thin foil, Thickness: 0.006 mm (0.0002 in)
Supplier: Thermo Scientific Chemicals
|
Warning
|
35813-FF
35813-FI
CAAA35813-FFEA
278.08
CAD
CAAA35813-FF
CAAA35813-FI
Titanium ≥99.6% (metals basis), thin foil, Thickness: 0.006 mm (0.0002 in)
Titanium
MDL: MFCD00011264
Purity: 99.6%
CAS No.: 7440-32-6
Physical Form: ≈0.02g/25x25mm, Not certified pinhole free
Odor: Odorless
MDL No.: MFCD00011264
Formula:
Ti Boiling Pt: 3262 °C (1013 hPa) Melting Pt: 1660 °C Density: 4.51 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 Merck Index: 12,09610 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...