Magnesium fluoride ≥99% (metals basis excluding Ca, Na) (max. 1% Ca + Na)
Supplier: Thermo Scientific Chemicals
|
Warning
|
35807-18
35807-30
35807-A1
CAAA35807-18EA
136.51
CAD
CAAA35807-18
CAAA35807-30
CAAA35807-A1
Magnesium fluoride ≥99% (metals basis excluding Ca, Na) (max. 1% Ca + Na)
Magnesium fluoride
MDL: MFCD00011108
Sparingly soluble in water. Slightly soluble in dilute HNO3
Purity: 99%
CAS No.: 7783-40-6
Molecular Formula: MgF2
Formula Weight: 62.30
Physical Form: -200 Mesh Powder
Density: 3.148
Refractive Index: 1.35-1.38
Appearance: White to off-white
Odor: Odorless
Melting Point: 1261°
Solubility: Sparingly soluble in water. Slightly soluble in dilute HNO3
Merck Reference: 14,5665
MDL No.: MFCD00011108
Formula:
MgF₂ Boiling Pt: 2260 °C (1013 hPa) Melting Pt: 1261 °C Density: 3.148 Storage Temperature: Ambient |
MDL Number:
MFCD00011108 CAS Number: 7783-40-6 EINECS: 231-995-1 Merck Index: 14,5665 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...