Samarium trichloride, ultra dry ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
35804-04
35804-09
35804-18
CAAA35804-04EA
98.59
CAD
CAAA35804-04
CAAA35804-09
CAAA35804-18
Samarium trichloride, ultra dry ≥99.9% (REO, rare earth oxide basis)
Samarium trichloride
MDL: MFCD00011235
Purity: 99.9%
CAS No.: 10361-82-7
Molecular Formula: SmCl3
Formula Weight: 256.71
Physical Form: Powder, Ampouled under argon
Density: 4.46
Melting Point: 678°
Boiling Point: dec.
Merck Reference: 14,8347
MDL No.: MFCD00011235
Formula:
Cl₃Sm Boiling Pt: Dec. Melting Pt: 678 °C Density: 4.46 Storage Temperature: Ambient |
MDL Number:
MFCD00011235 CAS Number: 10361-82-7 EINECS: 233-797-0 Merck Index: 14,8347 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...