Lutetium trichloride, ultra dry ≥99.98% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
35802-03
35802-06
CAAA35802-03EA
233.84
CAD
CAAA35802-03
CAAA35802-06
Lutetium trichloride, ultra dry ≥99.98% (REO, rare earth oxide basis)
Lutetium trichloride
MDL: MFCD00011099
Purity: 99.98%
CAS No.: 10099-66-8
Molecular Formula: LuCl3
Formula Weight: 281.33
Physical Form: Powder, Ampouled under argon
Density: 3.98
Melting Point: 905° subl.
Merck Reference: 14,5615
MDL No.: MFCD00011099
Formula:
Cl₃Lu Melting Pt: 905 °C (subl.) Density: 3.98 Storage Temperature: Ambient |
MDL Number:
MFCD00011099 CAS Number: 10099-66-8 EINECS: 233-240-1 Merck Index: 14,5615 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...