Lanthanum(III) iodide, ultra dry ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lanthanum triiodide

35801-03 35801-06
CAAA35801-03EA 159.58 CAD
CAAA35801-03 CAAA35801-06
Lanthanum(III) iodide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Lanthanum (III) iodide

MDL: MFCD00049470


Purity: 99.9% CAS No.: 13813-22-4 Molecular Formula: LaI3 Formula Weight: 519.62 Physical Form: -10 Mesh Beads, Ampouled under argon Sensitivity: Hygroscopic MDL No.: MFCD00049470

Formula: I₃La
MW: 519.62 g/mol
Melting Pt: 778
Density: 5.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049470
CAS Number: 13813-22-4
EINECS: 237-465-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR