Strontium carbonate ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
35793-14
35793-22
35793-36
CAAA35793-14EA
195.92
CAD
CAAA35793-14
CAAA35793-22
CAAA35793-36
Strontium carbonate ≥99.99% (metals basis)
Strontium carbonate
MDL: MFCD00011250
Notes: 1340°C -CO2
Purity: 99.99%
CAS No.: 1633-05-2
Molecular Formula: SrCO3
Formula Weight: 147.63
Physical Form: Powder
Density: 3.70
Melting Point: 1100° dec.
Merck Reference: 14,8838
MDL No.: MFCD00011250
Formula:
CO₃Sr Boiling Pt: 2647 °C (1 mmHg) Melting Pt: 1100 °C (dec.) Density: 3.70 Storage Temperature: Ambient |
MDL Number:
MFCD00011250 CAS Number: 1633-05-2 EINECS: 216-643-7 Merck Index: 14,8838 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...