Zinc acetate, anhydrous ≥99.98% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
35792-09
35792-18
CAAA35792-09EA
136.99
CAD
CAAA35792-09
CAAA35792-18
Zinc acetate, anhydrous ≥99.98% (metals basis)
Zinc acetate
MDL: MFCD00012454
Purity: 99.98%
CAS No.: 557-34-6
Molecular Formula: C4H6O4Zn
Formula Weight: 183.47
Physical Form: Granular
Density: 1.84
Appearance: White
Melting Point: 200° dec.
Merck Reference: 14,10128
UN No.: 3077
MDL No.: MFCD00012454
Formula:
C₄H₆O₄Zn MW: 183.48 g/mol Boiling Pt: 908 °C (1013 hPa) Melting Pt: 200 °C (dec.) Density: 1.84 Storage Temperature: Ambient |
MDL Number:
MFCD00012454 CAS Number: 557-34-6 EINECS: 209-170-2 UN: 3077 ADR: 9,III Merck Index: 14,10128 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...