Strontium ≥99% (metals basis), granules ≤ 19 mm (0.76 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Packaged under argon.
35789-06
35789-14
35789-22
CAAA35789-06EA
88.16
CAD
CAAA35789-06
CAAA35789-14
CAAA35789-22
Strontium ≥99% (metals basis), granules ≤ 19 mm (0.76 in)
Strontium
MDL: MFCD00134060
Purity: 99%
CAS No.: 7440-24-6
Physical Form: Packaged under argon with thin oil layer
Sensitivity: Air and Moisture Sensitive
Appearance: White to pale yellow
UN No.: 3208
MDL No.: MFCD00134060
Formula:
Sr Boiling Pt: 1151 °C (1013 hPa) Melting Pt: 570…660 °C Density: 2.38 g/cm³ (770 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134060 CAS Number: 7440-24-6 EINECS: 231-133-4 UN: 3208 ADR: 4.3,II Merck Index: 12,08994 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...