Sodium borohydride 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium tetrahydridoborate, Sodium tetrahydroborate
As a reducing agent for organic, inorganic and organometallic compounds, as a bleaching agent, in the recycling of gold and platinum metals, in organic synthesis, nonaqueous reducing agent for aldehydes, ketones, and schiff bases
Purity: 97+%
CAS No.: 16940-66-2
Molecular Formula: NaBH4
Formula Weight: 37.83
Physical Form: -10+40 Mesh Granules
Density: 1.074
Appearance: White
Odor: Weak
Melting Point: 400° dec.
Solubility: Soluble in water, ammonia and amines. Insoluble in ether
Merck Reference: 14,8592
UN No.: 1426
MDL No.: MFCD00003518
As a reducing agent for organic, inorganic and organometallic compounds, as a bleaching agent, in the recycling of gold and platinum metals, in organic synthesis, nonaqueous reducing agent for aldehydes, ketones, and schiff bases
Formula:
NaBH₄ Melting Pt: 400 °C (dec.) Density: 1.074 Flash Pt: 69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003518 CAS Number: 16940-66-2 EINECS: 241-004-4 UN: 1426 ADR: 4.3,I Merck Index: 14,8592 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...