Yttrium(III) iodide, ultra dry ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

35744-03 35744-06
CAAA35744-03EA 216.46 CAD
CAAA35744-03 CAAA35744-06
Yttrium(III) iodide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Yttrium (III) iodide

MDL: MFCD00054144


Purity: 99.9% CAS No.: 13470-38-7 Molecular Formula: YI3 Formula Weight: 469.62 Physical Form: Powder/Chunks, Ampouled under argon Sensitivity: Hygroscopic MDL No.: MFCD00054144

Formula: I₃Y
MW: 469.62
Melting Pt: 1004 °C
Storage Temperature: Ambient
MDL Number: MFCD00054144
CAS Number: 13470-38-7
EINECS: 236-737-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR