Yttrium(III) chloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
35743-09
35743-18
35743-30
CAAA35743-09EA
155.63
CAD
CAAA35743-09
CAAA35743-18
CAAA35743-30
Yttrium(III) chloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Yttrium (III) chloride
MDL: MFCD00011470
Purity: 99.99%
CAS No.: 10361-92-9
Molecular Formula: Cl3Y
Formula Weight: 195.26
Physical Form: Powder, Ampouled under argon
Density: 2.67
Appearance: White to pale yellow
Odor: Odorless
Melting Point: 721°
Merck Reference: 14,10107
MDL No.: MFCD00011470
Formula:
Cl₃Y Boiling Pt: 1488 °C (1013 hPa) Melting Pt: 721 °C Density: 2.67 Storage Temperature: Ambient |
MDL Number:
MFCD00011470 CAS Number: 10361-92-9 EINECS: 233-801-0 Merck Index: 14,10107 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...