Calcium fluoride, ultra dry ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

35742-06 35742-14 35742-22
CAAA35742-06EA 148.36 CAD
CAAA35742-06 CAAA35742-14 CAAA35742-22
Calcium fluoride, ultra dry ≥99.99% (metals basis)
Calcium fluoride

MDL: MFCD00010907


Purity: 99.99% CAS No.: 7789-75-5 Molecular Formula: CaF2 Formula Weight: 78.08 Physical Form: Powder, Ampouled under argon Density: 3.18 Refractive Index: 1.434 Sensitivity: Hygroscopic Odor: Odorless Melting Point: 1403° Boiling Point: ca 2500° Merck Reference: 14,1667 MDL No.: MFCD00010907

Formula: CaF₂
MW: 78.07 g/mol
Boiling Pt: ∼2500 °C
Melting Pt: 1403 °C
Density: 3.18
Storage Temperature: Ambient
MDL Number: MFCD00010907
CAS Number: 7789-75-5
EINECS: 232-188-7
Merck Index: 14,1667

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR