Cobalt(II) fluoride, ultra dry ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cobalt difluoride, Cobaltous fluoride
35740-04
35740-09
CAAA35740-04EA
87.22
CAD
CAAA35740-04
CAAA35740-09
Cobalt(II) fluoride, ultra dry ≥99.99% (metals basis)
Cobalt (II) fluoride
MDL: MFCD00010941
Purity: 99.99%
CAS No.: 10026-17-2
Molecular Formula: CoF2
Formula Weight: 96.93
Physical Form: Powder, Ampouled under argon
Density: 4.46
Melting Point: 1200°
Boiling Point: 1400°
Merck Reference: 14,2440
UN No.: 2923
MDL No.: MFCD00010941
Formula:
CoF₂ MW: 96.93 Boiling Pt: 1400 Melting Pt: 1200 Density: 4.46 Storage Temperature: Ambient |
MDL Number:
MFCD00010941 CAS Number: 10026-17-2 EINECS: 233-061-9 UN: 2923 ADR: 8 III Merck Index: 14,2440 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...