Erbium trichloride, ultra dry ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
35739-06
35739-14
CAAA35739-06EA
219.62
CAD
CAAA35739-06
CAAA35739-14
Erbium trichloride, ultra dry ≥99.9% (REO, rare earth oxide basis)
Erbium trichloride
MDL: MFCD00010989
Purity: 99.9%
CAS No.: 10138-41-7
Molecular Formula: ErCl3
Formula Weight: 273.62
Physical Form: Powder, Ampouled under argon
Sensitivity: Hygroscopic
Boiling Point: 1500°
MDL No.: MFCD00010989
Formula:
Cl₃Er Boiling Pt: 1500 °C Melting Pt: 774 °C Density: 4.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010989 CAS Number: 10138-41-7 EINECS: 233-385-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...