Europium(III) chloride, ultra dry ≥99.99% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

35738-03 35738-06 35738-14
CAAA35738-03EA 150.89 CAD
CAAA35738-03 CAAA35738-06 CAAA35738-14
Europium(III) chloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Europium (III) chloride

MDL: MFCD00010994 Soluble in water


Purity: 99.99% CAS No.: 10025-76-0 Molecular Formula: EuCl3 Formula Weight: 258.32 Physical Form: Powder, Ampouled under argon Sensitivity: Hygroscopic Solubility: Soluble in water Merck Reference: 14,3904 MDL No.: MFCD00010994

Formula: Cl₃Eu
Melting Pt: 850 °C
Density: 4.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010994
CAS Number: 10025-76-0
EINECS: 233-040-4
Merck Index: 14,3904

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR