Gadolinium tribromide, ultra dry ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Gadolinium (III) bromide

35737-04 35737-09
CAAA35737-04EA 77.26 CAD
CAAA35737-04 CAAA35737-09
Gadolinium tribromide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Gadolinium tribromide

MDL: MFCD00049741


Purity: 99.9% CAS No.: 13818-75-2 Molecular Formula: GdBr3 Formula Weight: 396.96 Physical Form: Powder, Ampouled under argon Sensitivity: Hygroscopic MDL No.: MFCD00049741

Formula: Br₃Gd
MW: 396.96 g/mol
Melting Pt: 770 °C
Storage Temperature: Ambient
MDL Number: MFCD00049741
CAS Number: 13818-75-2
EINECS: 237-494-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR