Gadolinium trichloride, ultra dry ≥99.99% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

35736-06 35736-14
CAAA35736-06EA 270.18 CAD
CAAA35736-06 CAAA35736-14
Gadolinium trichloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Gadolinium trichloride

MDL: MFCD00011024


Purity: 99.99% CAS No.: 10138-52-0 Molecular Formula: GdCl3 Formula Weight: 263.61 Physical Form: Powder, Ampouled Under Argon Sensitivity: Hygroscopic Appearance: White Melting Point: 609° Merck Reference: 14,4326 MDL No.: MFCD00011024

Formula: Cl₃Gd
MW: 263.61
Melting Pt: 609 °C
Density: 4.52
Storage Temperature: Ambient
MDL Number: MFCD00011024
CAS Number: 10138-52-0
EINECS: 233-386-6
Merck Index: 14,4326

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR