Gadolinium triiodide, ultra dry ≥99.99% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

35735-03 35735-06
CAAA35735-03EA 219.62 CAD
CAAA35735-03 CAAA35735-06
Gadolinium triiodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Gadolinium triiodide

MDL: MFCD00016114


Purity: 99.99% CAS No.: 13572-98-0 Molecular Formula: GdI3 Formula Weight: 537.96 Physical Form: Powder, Ampouled under argon Sensitivity: Hygroscopic MDL No.: MFCD00016114

Formula: GdI₃
Melting Pt: 926 °C
Storage Temperature: Ambient
MDL Number: MFCD00016114
CAS Number: 13572-98-0
EINECS: 236-997-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR