Lanthanum(III) chloride, ultra dry ≥99.99% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lanthanum chloride, Lanthanum trichloride

35731-06 35731-14 35731-22
CAAA35731-06EA 118.18 CAD
CAAA35731-06 CAAA35731-14 CAAA35731-22
Lanthanum(III) chloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Lanthanum (III) chloride

MDL: MFCD00011068


Purity: 99.99% CAS No.: 10099-58-8 Molecular Formula: LaCl3 Formula Weight: 245.27 Physical Form: Granular, Ampouled under argon Density: 3.842 Appearance: White Odor: Odorless Melting Point: 860° Boiling Point: 1000° Merck Reference: 14,5363 UN No.: 3077 MDL No.: MFCD00011068

Formula: Cl₃La
MW: 245.26 g/mol
Boiling Pt: 1000 °C
Melting Pt: 860 °C
Density: 3.842
Storage Temperature: Ambient
MDL Number: MFCD00011068
CAS Number: 10099-58-8
EINECS: 233-237-5
UN: 3077
ADR: 9,III
Merck Index: 14,5363

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR