Cobalt(II) iodide, ultra dry ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cobaltous iodide
35730-03
35730-06
CAAA35730-03EA
214.88
CAD
CAAA35730-03
CAAA35730-06
Cobalt(II) iodide, ultra dry ≥99.999% (metals basis)
Cobalt (II) iodide
MDL: MFCD00016030
Purity: 99.999%
CAS No.: 15238-00-3
Molecular Formula: CoI2
Formula Weight: 312.74
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 5.584
Appearance: White
Odor: Odorless
Melting Point: 515° vac.
Boiling Point: 570° vac.
Merck Reference: 14,2443
MDL No.: MFCD00016030
Formula:
CoI₂ MW: 312.74 g/mol Boiling Pt: 570 (in vacuum) Melting Pt: 515 Density: 5.584 Storage Temperature: Ambient |
MDL Number:
MFCD00016030 CAS Number: 15238-00-3 EINECS: 239-283-2 Merck Index: 14,2443 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...