Cesium iodide, dry_ultra 99,998 Metals basis
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cesiumiodide
35729-03
35729-09
35729-18
35729-30
CAAA35729-03EA
60.99
CAD
CAAA35729-03
CAAA35729-09
CAAA35729-18
CAAA35729-30
Cesium iodide, dry_ultra 99,998 Metals basis
Cesium iodide
MDL: MFCD00010962
Purity: 99.998%
CAS No.: 7789-17-5
Molecular Formula: CsI
Formula Weight: 259.81
Physical Form: -10 Mesh Beads, Ampouled under argon
Refractive Index: 1.7876
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 626°
Boiling Point: 1280°
Merck Reference: 14,2014
MDL No.: MFCD00010962
Formula:
CsI MW: 259.81 g/mol Boiling Pt: 1280 Melting Pt: 621 Density: 3.21 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010962 CAS Number: 7789-17-5 EINECS: 232-145-2 Merck Index: 14,2014 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...