Zinc chloride, ultra dry ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
35726-06
35726-14
35726-22
CAAA35726-06EA
109.34
CAD
CAAA35726-06
CAAA35726-14
CAAA35726-22
Zinc chloride, ultra dry ≥99.999% (metals basis)
Zinc chloride
MDL: MFCD00011295
Fieser: 1,1289 13,349 15,368 16,391 18,410 19,409 20,439 21,493
Purity: 99.999%
CAS No.: 7646-85-7
Molecular Formula: Cl2Zn
Formula Weight: 136.29
Physical Form: Powder, Ampouled under argon
Density: 2.91
Melting Point: 290°
Boiling Point: 732°
Merck Reference: 14,10132
UN No.: 2331
MDL No.: MFCD00011295
Formula:
ZnCl₂ Boiling Pt: 732 °C Melting Pt: 290 °C Density: 2.91 Storage Temperature: Ambient |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2331 ADR: 8,III Merck Index: 14,10132 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...