Tin(II) chloride, ultra dry ≥99.9985% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
35723-03
35723-06
35723-14
35723-22
CAAA35723-03EA
109.49
CAD
CAAA35723-03
CAAA35723-06
CAAA35723-14
CAAA35723-22
Tin(II) chloride, ultra dry ≥99.9985% (metals basis)
Tin (II) chloride
MDL: MFCD00011241
Fieser: 1,1113 15,309 16,329 17,334 18,353 19,337 20,373 21,422
Purity: 99.9985%
CAS No.: 7772-99-8
Molecular Formula: SnCl2
Formula Weight: 189.62
Physical Form: Powder, Ampouled under argon
Density: 3.95
Melting Point: 247°
Boiling Point: 652°
Merck Reference: 14,8783
UN No.: 3260
MDL No.: MFCD00011241
Formula:
SnCl₂ Boiling Pt: 652 °C Melting Pt: 247 °C Density: 3.95 Storage Temperature: Ambient |
MDL Number:
MFCD00011241 CAS Number: 7772-99-8 EINECS: 231-868-0 UN: 3260 ADR: 8,III Merck Index: 14,8783 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...