Thallium(I) iodide, ultra dry ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
35722-09
35722-18
35722-30
CAAA35722-09EA
102.07
CAD
CAAA35722-09
CAAA35722-18
CAAA35722-30
Thallium(I) iodide, ultra dry ≥99.999% (metals basis)
Thallium (I) iodide
MDL: MFCD00011279
Purity: 99.999%
CAS No.: 7790-30-9
Molecular Formula: TlI
Formula Weight: 331.27
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 7.1
Appearance: Yellow
Odor: Odorless
Melting Point: 440°
Boiling Point: 824°
Merck Reference: 14,9264
UN No.: 1707
MDL No.: MFCD00011279
Formula:
ITl Boiling Pt: 824 °C Melting Pt: 440 °C Density: 7.1 Storage Temperature: Ambient |
MDL Number:
MFCD00011279 CAS Number: 7790-30-9 EINECS: 232-199-7 UN: 1707 ADR: 6.1,II Merck Index: 14,9264 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...