Thallium(I) chloride, ultra dry ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

35721-06 35721-14
CAAA35721-06EA 230.68 CAD
CAAA35721-06 CAAA35721-14
Thallium(I) chloride, ultra dry ≥99.999% (metals basis)
Thallium (I) chloride

MDL: MFCD00011274


Purity: 99.999% CAS No.: 7791-12-0 Molecular Formula: TlCl Formula Weight: 239.82 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 7.0 Refractive Index: 2.247 Sensitivity: Air Sensitive Melting Point: 430° Merck Reference: 14,9260 UN No.: 1707 MDL No.: MFCD00011274

Formula: ClTl
Boiling Pt: 720 °C (1013 hPa)
Melting Pt: 430 °C
Density: 7.0
Storage Temperature: Ambient
MDL Number: MFCD00011274
CAS Number: 7791-12-0
EINECS: 232-241-4
UN: 1707
ADR: 6.1,II
Merck Index: 14,9260

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR