Strontium bromide, ultra dry ≥99.995% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

35717-03 35717-06
CAAA35717-03EA 128.61 CAD
CAAA35717-03 CAAA35717-06
Strontium bromide, ultra dry ≥99.995% (metals basis)
Strontium bromide

MDL: MFCD00049548


Purity: 99.995% CAS No.: 10476-81-0 Molecular Formula: BrSr2 Formula Weight: 247.44 Physical Form: Powder, Ampouled under argon Density: 4.216 Appearance: White Odor: Odorless Melting Point: 643° Merck Reference: 14,8837 MDL No.: MFCD00049548

Formula: Br₂Sr
MW: 247.43 g/mol
Boiling Pt: 2146 °C (1013 hPa)
Melting Pt: 643 °C
Density: 4.216
Storage Temperature: Ambient
MDL Number: MFCD00049548
CAS Number: 10476-81-0
EINECS: 233-969-5
Merck Index: 14,8837

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR