Sodium chloride, ultra dry ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
Warning
|
35716-06
35716-14
35716-22
CAAAA35716-06EA
85.79
CAD
CAAAA35716-06
CAAA35716-14
CAAA35716-22
Sodium chloride, ultra dry ≥99.99% (metals basis)
Sodium chloride
MDL: MFCD00003477
Fieser: 6,285
Purity: 99.99%
CAS No.: 7647-14-5
Molecular Formula: NaCl
Formula Weight: 58.44
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 2.165
Refractive Index: 1.5442
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
pH: 6-9
Melting Point: 801°
Boiling Point: 1413°
Merck Reference: 14,8599
MDL No.: MFCD00003477
Formula:
NaCl Boiling Pt: 1413 °C Melting Pt: 801 °C Density: 2.165 Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,8599 |
Specification Test Results
Form | -10 Mesh Beads, Ampouled under argon |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...