Praseodymium tribromide, ultra dry ≥99.99% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

35714-04 35714-09
CAAA35714-04EA 325.48 CAD
CAAA35714-04 CAAA35714-09
Praseodymium tribromide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Praseodymium tribromide

MDL: MFCD00049762


Purity: 99.99% CAS No.: 13536-53-3 Molecular Formula: PrBr3 Formula Weight: 380.63 Physical Form: Powder, Ampouled under argon Sensitivity: Hygroscopic MDL No.: MFCD00049762

Formula: Br₃Pr
MW: 380.62 g/mol
Melting Pt: 691 °C
Storage Temperature: Ambient
MDL Number: MFCD00049762
CAS Number: 13536-53-3
EINECS: 236-893-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR