Nickel(II) chloride, ultra dry ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Nickel dichloride
35713-06
35713-14
35713-22
CAAA35713-06EA
214.88
CAD
CAAA35713-06
CAAA35713-14
CAAA35713-22
Nickel(II) chloride, ultra dry ≥99.9% (metals basis)
Nickel (II) chloride
MDL: MFCD00011142
Fieser: 12,336 18,249 20,258 21,281
Purity: 99.9%
CAS No.: 7718-54-9
Molecular Formula: NiCl2
Formula Weight: 129.60
Physical Form: Powder, Ampouled under argon
Density: 3.55
Appearance: Orange
Odor: Odorless
Melting Point: 1001°
Merck Reference: 14,6505
UN No.: 3288
MDL No.: MFCD00011142
Formula:
Cl₂Ni Melting Pt: 1001 °C Density: 3.55 Storage Temperature: Ambient |
MDL Number:
MFCD00011142 CAS Number: 7718-54-9 EINECS: 231-743-0 UN: 3288 ADR: 6.1,III Merck Index: 14,6505 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...