Manganese(II) iodide, ultra dry ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
35710-04
35710-09
CAAA35710-04EA
254.38
CAD
CAAA35710-04
CAAA35710-09
Manganese(II) iodide, ultra dry ≥99.99% (metals basis)
Manganese (II) iodide
MDL: MFCD00016225
Purity: 99.99%
CAS No.: 7790-33-2
Molecular Formula: I2Mn
Formula Weight: 308.75
Physical Form: Powder/Flakes, Ampouled under argon
Density: 5.0
Appearance: White to pink to red
Odor: Odorless
Melting Point: 638°
Merck Reference: 14,5732
MDL No.: MFCD00016225
Formula:
I₂Mn Melting Pt: 638 °C Density: 5.0 Storage Temperature: Ambient |
MDL Number:
MFCD00016225 CAS Number: 7790-33-2 EINECS: 232-201-6 Merck Index: 14,5732 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...