Magnesium chloride, ultra dry ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
Warning
|
35707-09
35707-18
CAAA35707-09EA
123.71
CAD
CAAA35707-09
CAAA35707-18
Magnesium chloride, ultra dry ≥99.9% (metals basis)
Magnesium chloride
MDL: MFCD00011106
Purity: 99.9%
CAS No.: 7786-30-3
Molecular Formula: Cl2Mg
Formula Weight: 95.22
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 2.32
Appearance: White to yellow
Odor: Odorless
Melting Point: 714°
Merck Reference: 14,5662
MDL No.: MFCD00011106
Formula:
MgCl₂ Boiling Pt: 1412 °C (1013 hPa) Melting Pt: 714 °C Density: 2.32 Storage Temperature: Ambient |
MDL Number:
MFCD00011106 CAS Number: 7786-30-3 EINECS: 232-094-6 Merck Index: 14,5662 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...