Lithium bromide, ultra dry ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
35705-14
35705-22
CAAA35705-14EA
199.08
CAD
CAAA35705-14
CAAA35705-22
Lithium bromide, ultra dry ≥99.9% (metals basis)
Lithium bromide
MDL: MFCD00011077
Purity: 99.9%
CAS No.: 7550-35-8
Molecular Formula: BrLi
Formula Weight: 86.85
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 3.464
Appearance: White
Odor: Odorless
Melting Point: 550°
Boiling Point: 1265°
Merck Reference: 14,5526
MDL No.: MFCD00011077
Formula:
LiBr Boiling Pt: 1265 °C Melting Pt: 550 °C Density: 3.464 Storage Temperature: Ambient |
MDL Number:
MFCD00011077 CAS Number: 7550-35-8 EINECS: 231-439-8 Merck Index: 14,5526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...