Lead(II) bromide, ultra dry ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead dibromide

35703-03 35703-06 35703-14
CAAA35703-03EA 128.3 CAD
CAAA35703-03 CAAA35703-06 CAAA35703-14
Lead(II) bromide, ultra dry ≥99.999% (metals basis)
Lead (II) bromide

MDL: MFCD00011156


Purity: 99.999% CAS No.: 10031-22-8 Molecular Formula: Br2Pb Formula Weight: 367.01 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 6.66 Appearance: White Odor: Odorless Melting Point: 373° Boiling Point: 916° Merck Reference: 14,5403 UN No.: 2291 MDL No.: MFCD00011156

Formula: Br₂Pb
MW: 367.01 g/mol
Boiling Pt: 916 °C
Melting Pt: 373 °C
Density: 6.66
Storage Temperature: Ambient
MDL Number: MFCD00011156
CAS Number: 10031-22-8
EINECS: 233-084-4
UN: 2291
ADR: 6.1,III
Merck Index: 14,5403

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR