Gallium tribromide, ultra dry ≥99.998% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
35697-03
35697-06
CAAA35697-03EA
181.7
CAD
CAAA35697-03
CAAA35697-06
Gallium tribromide, ultra dry ≥99.998% (metals basis)
Gallium tribromide
MDL: MFCD00016100
Purity: 99.998%
CAS No.: 13450-88-9
Molecular Formula: GaBr3
Formula Weight: 309.45
Physical Form: Crystalline, Ampouled under argon
Density: 3.69
Appearance: White
Odor: Odorless
Melting Point: 122°
Boiling Point: 279°
UN No.: 3260
MDL No.: MFCD00016100
Formula:
Br₃Ga Boiling Pt: 279 °C Melting Pt: 122 °C Density: 3.69 Storage Temperature: Ambient |
MDL Number:
MFCD00016100 CAS Number: 13450-88-9 EINECS: 236-609-5 UN: 3260 ADR: 8,II |
Specification Test Results
Form | Crystalline, Ampouled under argon |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...