Erbium triiodide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
35695-03
35695-06
CAAA35695-03EA
143.31
CAD
CAAA35695-03
CAAA35695-06
Erbium triiodide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Erbium triiodide
MDL: MFCD00016075
Purity: 99.9%
CAS No.: 13813-42-8
Molecular Formula: ErI3
Formula Weight: 547.97
Physical Form: Pieces/Powder, Ampouled under argon
Sensitivity: Hygroscopic
Boiling Point: 1280°
MDL No.: MFCD00016075
Formula:
ErI₃ Boiling Pt: 1280 °C Melting Pt: 1020 °C Density: ∼5.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016075 CAS Number: 13813-42-8 EINECS: 237-471-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...