Dysprosium triiodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
35693-03
35693-06
CAAA35693-03EA
178.54
CAD
CAAA35693-03
CAAA35693-06
CAAA35693-14
Dysprosium triiodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Dysprosium triiodide
MDL: MFCD00016072
Purity: 99.99%
CAS No.: 15474-63-2
Molecular Formula: DyI3
Formula Weight: 543.21
Physical Form: Flake, Ampouled under argon
Sensitivity: Hygroscopic
Appearance: Green to yellow
Melting Point: 955°
MDL No.: MFCD00016072
Formula:
DyI₃ Melting Pt: 955 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016072 CAS Number: 15474-63-2 EINECS: 239-493-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...