Cerium trichloride, ultra dry ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
35688-22
35688-06
35688-14
CAAAA35688-22EA
1199.22
CAD
CAAAA35688-22
CAAA35688-06
CAAA35688-14
Cerium trichloride, ultra dry ≥99.9% (REO, rare earth oxide basis)
Cerium trichloride
MDL: MFCD00010929
Purity: 99.9%
CAS No.: 7790-86-5
Molecular Formula: CeCl3
Formula Weight: 246.48
Physical Form: Crystalline, Ampouled under argon
Density: 3.92
Appearance: White to pale grey
Odor: Odorless
Melting Point: 848°
Boiling Point: 1727°
Merck Reference: 14,1997
MDL No.: MFCD00010929
Formula:
CeCl₃ Boiling Pt: 1727 °C Melting Pt: 848 °C Density: 3.92 Storage Temperature: Ambient |
MDL Number:
MFCD00010929 CAS Number: 7790-86-5 EINECS: 232-227-8 Merck Index: 14,1997 |
Specification Test Results
Form | Crystalline, Ampouled under argon |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...