Calcium chloride, ultra dry ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

35686-03 35686-09
CAAA35686-03EA 75.05 CAD
CAAA35686-03 CAAA35686-09
Calcium chloride, ultra dry ≥99.99% (metals basis)
Calcium chloride

MDL: MFCD00010903


Purity: 99.99% CAS No.: 10043-52-4 Molecular Formula: CaCl2 Formula Weight: 110.99 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 2.15 Appearance: White Melting Point: 782° Boiling Point: 1600° Merck Reference: 14,1659 MDL No.: MFCD00010903

Formula: CaCl₂
Boiling Pt: >1600 °C
Melting Pt: 782 °C
Density: 2.15
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,1659

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR