Calcium chloride, ultra dry ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
35685-04
35685-09
CAAA35685-04EA
208.56
CAD
CAAA35685-04
CAAA35685-09
Calcium chloride, ultra dry ≥99.9% (metals basis)
Calcium chloride
MDL: MFCD00010903
Purity: 99.9%
CAS No.: 10043-52-4
Molecular Formula: CaCl2
Formula Weight: 110.99
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 2.15
Appearance: White
Melting Point: 782°
Boiling Point:
Formula:
CaCl₂ Boiling Pt: >1600 °C Melting Pt: 782 °C Density: 2.15 Storage Temperature: Ambient |
MDL Number:
MFCD00010903 CAS Number: 10043-52-4 EINECS: 233-140-8 REACH: 01-2119494219-28 Merck Index: 14,1659 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...