Calcium bromide, ultra dry ≥99.978% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
35684-03
35684-06
CAAA35684-03EA
71.57
CAD
CAAA35684-03
CAAA35684-06
Calcium bromide, ultra dry ≥99.978% (metals basis)
Calcium bromide
MDL: MFCD00010902
Purity: 99.978%
CAS No.: 7789-41-5
Molecular Formula: Br2Ca
Formula Weight: 199.90
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 3.353
Appearance: White to off-white
Odor: Odorless
Melting Point: 730°
Boiling Point: 806-812°
Merck Reference: 14,1654
MDL No.: MFCD00010902
Formula:
Br₂Ca Boiling Pt: 806…812 °C Melting Pt: 730 °C Density: 3.353 Storage Temperature: Ambient |
MDL Number:
MFCD00010902 CAS Number: 7789-41-5 EINECS: 232-164-6 Merck Index: 14,1654 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...