Cadmium(II) iodide, ultra dry ≥99.9985% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
35683-03
35683-06
35683-14
CAAA35683-03EA
46.45
CAD
CAAA35683-03
CAAA35683-06
CAAA35683-14
Cadmium(II) iodide, ultra dry ≥99.9985% (metals basis)
Cadmium (II) iodide
MDL: MFCD00010920
Purity: 99.9985%
CAS No.: 7790-80-9
Molecular Formula: CdI2
Formula Weight: 366.21
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 5.67
Appearance: White to yellow
Odor: Odorless
Melting Point: 387-389°
Boiling Point: 787°
Merck Reference: 14,1621
UN No.: 2570
MDL No.: MFCD00010920
Formula:
CdI₂ Boiling Pt: 787 °C Melting Pt: 387…389 °C Density: 5.67 Storage Temperature: Ambient |
MDL Number:
MFCD00010920 CAS Number: 7790-80-9 EINECS: 232-223-6 UN: 2570 ADR: 6.1,III Merck Index: 14,1621 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...