Cadmium(II) bromide, ultra dry ≥99.999%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cadmium bromide
35682-06
35682-14
CAAA35682-06EA
202.24
CAD
CAAA35682-06
CAAA35682-14
Cadmium(II) bromide, ultra dry ≥99.999%
Cadmium (II) bromide
MDL: MFCD00015996
Purity: 99.999%
CAS No.: 7789-42-6
Molecular Formula: CdBr2
Formula Weight: 272.22
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 5.192
Appearance: White to pale yellow
Odor: Odorless
Melting Point: 567°
Boiling Point: 863°
Merck Reference: 14,1615
UN No.: 2570
MDL No.: MFCD00015996
Formula:
Br₂Cd MW: 272.22 g/mol Boiling Pt: 863 Melting Pt: 567 Density: 5.192 Storage Temperature: Ambient |
MDL Number:
MFCD00015996 CAS Number: 7789-42-6 EINECS: 232-165-1 UN: 2570 ADR: 6.1 III Merck Index: 14,1615 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...