Bismuth(III) iodide, ultra dry ≥99.998% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
35681-06
35681-14
CAAA35681-06EA
259.12
CAD
CAAA35681-06
CAAA35681-14
Bismuth(III) iodide, ultra dry ≥99.998% (metals basis)
Bismuth (III) iodide
MDL: MFCD00010894
Purity: 99.998%
CAS No.: 7787-64-6
Molecular Formula: BiI3
Formula Weight: 589.69
Physical Form: -10 Mesh Granules, Ampouled under argon
Density: 5.778
Melting Point: 408°
Boiling Point: ca 500°
Merck Reference: 14,1269
UN No.: 3260
MDL No.: MFCD00010894
Formula:
BiI₃ Boiling Pt: ∼500 °C Melting Pt: 408 °C Density: 5.778 Storage Temperature: Ambient |
MDL Number:
MFCD00010894 CAS Number: 7787-64-6 EINECS: 232-127-4 UN: 3260 ADR: 8,III Merck Index: 14,1269 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...