Barium iodide, ultra dry ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
35679-06
35679-14
CAAA35679-06EA
287.56
CAD
CAAA35679-06
CAAA35679-14
Barium iodide, ultra dry ≥99.995% (metals basis)
Barium iodide
Barium iodide is used to prepare organobarium compounds by reacting with alkyl potassium compounds. It is also involved in the preparation of active barium metal by reducing with lithium biphenyl.
Purity: 99.995%
CAS No.: 13718-50-8
Molecular Formula: BaI2
Formula Weight: 391.15
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 5.15
Appearance: White
Odor: Odorless
Melting Point: 740°
Merck Reference: 14,980
UN No.: 1564
MDL No.: MFCD00003452
Formula:
BaI₂ Melting Pt: 740 °C Density: 5.15 Storage Temperature: Ambient |
MDL Number:
MFCD00003452 CAS Number: 13718-50-8 EINECS: 237-276-9 UN: 1564 ADR: 6.1,III Merck Index: 14,980 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...