Barium bromide, ultra dry ≥99.998% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

35678-03 35678-06
CAAA35678-03EA 101.28 CAD
CAAA35678-03 CAAA35678-06
Barium bromide, ultra dry ≥99.998% (metals basis)
Barium bromide

MDL: MFCD00003444


Purity: 99.998% CAS No.: 10553-31-8 Molecular Formula: BaBr2 Formula Weight: 297.14 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 4.781 Appearance: White Odor: Odorless Melting Point: 850° Merck Reference: 14,968 UN No.: 1564 MDL No.: MFCD00003444

Formula: BaBr₂
MW: 297.14 g/mol
Boiling Pt: 1835 °C (1013 hPa)
Melting Pt: 850 °C
Density: 4.781
Storage Temperature: Ambient
MDL Number: MFCD00003444
CAS Number: 10553-31-8
EINECS: 234-140-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,968

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR